2,4-Quinolinediol ≥97.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
D1753-500G
D1753-25G
TCD1753-500GEA
537.2
CAD
TCD1753-500G
TCD1753-025G
2,4-Quinolinediol ≥97.0% (by GC, titration analysis)
2,4-Quinolinediol
CAS Number: 86-95-3
MDL Number: MFCD00006744
Molecular Formula: C9H7NO2
Molecular Weight: 161.16
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Other Product Names / Synonyms: 2,4-Quinolinediol, 4-Hydroxy-2-quinolinone
Formula:
C₉H₇NO₂ Boiling Pt: 375 °C (1013 hPa) Melting Pt: 159 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006744 CAS Number: 86-95-3 EINECS: 201-711-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...