(±)-1,4-Dibromo-2-butanol ≥95.0%

Supplier: TCI America

Synonyms: 1,4-Dibromo-2-butanol

D1743-5G D1743-25G
TCD1743-005GEA 50.56 CAD
TCD1743-005G TCD1743-025G
(±)-1,4-Dibromo-2-butanol ≥95.0%
(±)-1,4-Dibromo-2-butanol

CAS Number: 19398-47-1
MDL Number: MFCD00000252
Molecular Formula: C4H8Br2O
Molecular Weight: 231.92
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 118
Flash Point (°C): 113
Specific Gravity (20/20): 1.97

Formula: BrH₂CCH(OH)CH₂CH₂Br
MW: 231.91 g/mol
Boiling Pt: 118 °C
Density: 1.97 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00000252
CAS Number: 19398-47-1
EINECS: 243-029-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04de7a3cc53e9c49b7e2ec1be5fb1c023f15afc7&vid=04de7a3cc53e9c49b7e2ec1be5fb1c023f15afc7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR