2,5-Dichloroterephthalonitrile ≥99.0%
Supplier: TCI America
Synonyms:
1,4-Dichloro-2,5-dicyanobenzene
D1734-5G
D1734-1G
TCD1734-005GEA
1423.58
CAD
TCD1734-005G
TCD1734-001G
2,5-Dichloroterephthalonitrile ≥99.0%
2,5-Dichloroterephthalonitrile
CAS Number: 1897-43-4
MDL Number: MFCD00059584
Molecular Formula: C8H2Cl2N2
Molecular Weight: 197.02
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 214
Other Product Names / Synonyms: 1,4-Dichloro-2,5-dicyanobenzene
Formula:
C₈H₂Cl₂N₂ MW: 197.02 g/mol Melting Pt: 214 °C |
MDL Number:
MFCD00059584 CAS Number: 1897-43-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...