5,5-Dibromo-2,2-dimethyl-1,3-dioxane-4,6-dione (5,5-Dibromomeldrum's acid) ≥98.0%

Supplier: TCI America

Synonyms: 5,5-Dibromo-2,2-dimethyl-1,3-dioxane-4,6-dione, cycl-Isopropylidene dibromomalonate, 5,5-Dibromomeldrum's acid (=5,5-Dibromo-2,2-dimethyl-4,6-dioxy-1,3-dioxane), Dibromomalonic acid Cyclic Isopropylidine ester, 5,5-Dibromo-2,2-dimethyl-4,6-dioxo-1,3-dioxane

D1710-25G D1710-5G
TCD1710-25GEA 791.74 CAD
TCD1710-25G TCD1710-005G
5,5-Dibromo-2,2-dimethyl-1,3-dioxane-4,6-dione (5,5-Dibromomeldrum's acid) ≥98.0%
5,5-Dibromo-2,2-dimethyl-1,3-dioxane-4,6-dione (5,5-Dibromomeldrum's acid)

CAS Number: 66131-14-4
MDL Number: MFCD00059808
Molecular Formula: C6H6Br2O4
Molecular Weight: 301.92
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 75
Storage Temperature: <0°C


Product Categories: Synthetic Organic Chemistry, Halogenation
Other Product Names / Synonyms: 5,5-Dibromo-2,2-dimethyl-4,6-dioxo-1,3-dioxane, Dibromomalonic Acid Cyclic Isopropylidine Ester, cycl-Isopropylidene Dibromomalonate

Formula: C₆H₆Br₂O₄
MW: 301.9174 g/mol
Melting Pt: 75 °C
MDL Number: MFCD00059808
CAS Number: 66131-14-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e5903e18522a6fc0d190379025e7fd2213a672b&vid=5e5903e18522a6fc0d190379025e7fd2213a672b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR