Di(2-pyridyl) ketone oxime ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D1709-1G D1709-5G
TCD1709-001GEA 74.42 CAD
TCD1709-001G TCD1709-005G
Di(2-pyridyl) ketone oxime ≥98.0% (by HPLC, titration analysis)
Di(2-pyridyl) ketone oxime

CAS Number: 1562-95-4
MDL Number: MFCD00006311
Molecular Formula: C11H9N3O
Molecular Weight: 199.21
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 142

Formula: C₁₁H₉N₃O
Melting Pt: 142 °C
MDL Number: MFCD00006311
CAS Number: 1562-95-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0ae5e039d70e22f8a08ec01d93e7c7cd9d8cb68&vid=f0ae5e039d70e22f8a08ec01d93e7c7cd9d8cb68&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR