(S)-(-)-N,N-Dimethyl-1-phenylethylamine ≥98.0%

Supplier: TCI America

Synonyms: (S)-(-)-alpha-Methyl-N,N-dimethylbenzylamine, (S)-N,N-Dimethyl-1-phenylethanamine

D1687-1G D1687-5G
TCD1687-001GEA 118.97 CAD
TCD1687-001G TCD1687-005G
(S)-(-)-N,N-Dimethyl-1-phenylethylamine ≥98.0%
(S)-(-)-N,N-Dimethyl-1-phenylethylamine

CAS Number: 17279-31-1
MDL Number: MFCD00056236
Molecular Formula: C10H15N
Molecular Weight: 149.24
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 71
Flash Point (°C): 64
Specific Gravity (20/20): 0.91
Specific rotation [a]20/D: -70 deg (neat)


Other Product Names / Synonyms: (S)-(-)-alpha-Methyl-N,N-dimethylbenzylamine

Formula: C₁₀H₁₅N
MW: 149.24 g/mol
Boiling Pt: 71 °C
Density: 0.91 (20 °C)
Flash Pt: 64 °C
MDL Number: MFCD00056236
CAS Number: 17279-31-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b31809da7d9f16d0cb9448bf5cc126b5f38356d&vid=9b31809da7d9f16d0cb9448bf5cc126b5f38356d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR