3,4'-Methylenedianiline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
D1684-25G
D1684-5G
D1684-1G
TCD1684-25GEA
840.56
CAD
TCD1684-25G
TCD1684-5G
TCD1684-001G
3,4'-Methylenedianiline ≥98.0% (by GC, titration analysis)
3,4'-Methylenedianiline
CAS Number: 19430-83-2
MDL Number: MFCD00014778
Molecular Formula: C13H14N2
Molecular Weight: 198.27
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 86
Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials, Diphenylmethanes (for High-Performance Polymer Research)
Other Product Names / Synonyms: 3,4'-Methylenedianiline
Formula:
C₁₃H₁₄N₂ Melting Pt: 86 °C |
MDL Number:
MFCD00014778 CAS Number: 19430-83-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...