2-Diethylaminopyridine ≥97.0% (by titrimetric analysis)
Supplier: TCI America
D1681-25G
D1681-5G
TCD1681-025GEA
424.86
CAD
TCD1681-025G
TCD1681-5G
2-Diethylaminopyridine ≥97.0% (by titrimetric analysis)
2-Diethylaminopyridine
CAS Number: 36075-06-6
MDL Number: MFCD00059777
Molecular Formula: C9H14N2
Molecular Weight: 150.23
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Boiling point (°C): 118
Specific Gravity (20/20): 0.97
Other Product Names / Synonyms: N-(2-Pyridyl)diethylamine
Formula:
C₉H₁₄N₂ Boiling Pt: 118 °C Density: 0.97 (20 °C) |
MDL Number:
MFCD00059777 CAS Number: 36075-06-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...