5-Amino-8-hydroxyquinoline dihydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
5-Amino-8-quinolinol Dihydrochloride, 5-Amino-8-hydroxyquinoline Dihydrochloride
A1005-25G
A1005-5G
TCA1005-25GEA
838.03
CAD
TCA1005-25G
TCA1005-005G
5-Amino-8-hydroxyquinoline dihydrochloride ≥98.0%
5-Amino-8-hydroxyquinoline dihydrochloride
CAS Number: 21302-43-2
MDL Number: MFCD00012737
Molecular Formula: C9H8N2O
Molecular Weight: 233.09
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: 5-Amino-8-quinolinol Dihydrochloride
Formula:
C₉H₈N₂O MW: 233.09 g/mol |
MDL Number:
MFCD00012737 CAS Number: 21302-43-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...