4-Dimethylamino-1-naphthyl isothiocyanate ≥98.0%

Supplier: TCI America

Synonyms: DANITO

D1669-1G D1669-5G
TCD1669-001GEA 237 CAD
TCD1669-001G TCD1669-005G
4-Dimethylamino-1-naphthyl isothiocyanate ≥98.0%
4-Dimethylamino-1-naphthyl isothiocyanate

[for HPLC Labeling]
CAS Number: 29711-79-3
MDL Number: MFCD00003883
Molecular Formula: C13H12N2S
Molecular Weight: 228.31
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 67


Product Categories: Analytical Science, Fluorescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: DANITO

Formula: C₁₃H₁₂N₂S
MW: 228.31 g/mol
Melting Pt: 67 °C
MDL Number: MFCD00003883
CAS Number: 29711-79-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=595407c25fd750d24d88eddd248d184a19d31118&vid=595407c25fd750d24d88eddd248d184a19d31118&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR