2,3-Dimethylcyclohexanol (mixture of isomers) ≥98.0% for synthesis
Supplier: TCI America
Synonyms:
2,3-Dimethylcyclohexanol
D1660-25ML
D1660-5ML
TCD1660-025MLEA
420.28
CAD
TCD1660-025ML
TCD1660-5ML
2,3-Dimethylcyclohexanol (mixture of isomers) ≥98.0% for synthesis
2,3-Dimethylcyclohexanol (mixture of isomers)
CAS Number: 1502-24-5
MDL Number: MFCD00001444
Molecular Formula: C8H16O
Molecular Weight: 128.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 181
Flash Point (°C): 66
Specific Gravity (20/20): 0.93
Formula:
C₈H₁₆O MW: 128.21 g/mol Boiling Pt: 181 °C Density: 0.93 (20 °C) Flash Pt: 66 °C |
MDL Number:
MFCD00001444 CAS Number: 1502-24-5 EINECS: 216-121-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...