Stearyl-3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate ≥98.0%

Supplier: TCI America

Synonyms: Octadecyl 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionate, 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic Acid Stearyl Ester

D1644-500G D1644-25G D1644-100G
TCD1644-500GEA 297.04 CAD
TCD1644-500G TCD1644-025G TCD1644-100G
Stearyl-3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate ≥98.0%
Stearyl-3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate

CAS Number: 2082-79-3
MDL Number: MFCD00017498
Molecular Formula: C35H62O3
Molecular Weight: 530.88
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 53
Flash Point (°C): 110


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic Acid Stearyl Ester, Octadecyl 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionate

Formula: C₃₅H₆₂O₃
MW: 530.88 g/mol
Melting Pt: 53 °C
Flash Pt: 110 °C
MDL Number: MFCD00017498
CAS Number: 2082-79-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24b26b95436a3822fec3e0e773f312dc1d6450ab&vid=24b26b95436a3822fec3e0e773f312dc1d6450ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR