4,4'-Diaminooctafluorobiphenyl ≥97.0%

Supplier: TCI America

Synonyms: 2,2',3,3',5,5',6,6'-Octafluoro-[1,1'-biphenyl]-4,4'-diamine, Octafluorobenzidine

D1632-5G D1632-1G
TCD1632-005GEA 173.8 CAD
TCD1632-005G TCD1632-1G
4,4'-Diaminooctafluorobiphenyl ≥97.0%
4,4'-Diaminooctafluorobiphenyl

CAS Number: 1038-66-0
MDL Number: MFCD00007646
Molecular Formula: C12H4F8N2
Molecular Weight: 328.17
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 177


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Materials Chemistry, Monomers, Polyimide Raw Materials, Biphenyls (for High-Performance Polymer Research)
Other Product Names / Synonyms: Octafluorobenzidine

Formula: C₁₂H₄F₈N₂
MW: 328.17 g/mol
Melting Pt: 177 °C
MDL Number: MFCD00007646
CAS Number: 1038-66-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddf68f0dbcab577356361e22cbb390f90eab6906&vid=ddf68f0dbcab577356361e22cbb390f90eab6906&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR