4-Dimethylaminobenzylamine dihydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 4-(Aminomethyl)-N,N-dimethylaniline dihydrochloride

D1620-5G D1620-25G
TCD1620-005GEA 335.59 CAD
TCD1620-005G TCD1620-025G
4-Dimethylaminobenzylamine dihydrochloride ≥98.0%
4-Dimethylaminobenzylamine dihydrochloride

CAS Number: 34403-52-6
MDL Number: MFCD00012862
Molecular Formula: C9H14N2
Molecular Weight: 223.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₉H₁₄N₂
MDL Number: MFCD00012862
CAS Number: 34403-52-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b529d97ce89b4c589e9e39380cb2ba6177c684ca&vid=b529d97ce89b4c589e9e39380cb2ba6177c684ca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR