5,6,7,8-Tetrahydronaphthalen-1-amine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A0995-25ML A0995-5ML
TCA0995-025MLEA 423.44 CAD
TCA0995-025ML TCA0995-005ML
5,6,7,8-Tetrahydronaphthalen-1-amine ≥98.0% (by GC, titration analysis)
5,6,7,8-Tetrahydronaphthalen-1-amine

CAS Number: 2217-41-6
MDL Number: MFCD00001736
Molecular Formula: C10H13N
Molecular Weight: 147.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 275
Flash Point (°C): 71
Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: 1-Amino-5,6,7,8-tetrahydronaphthalene, 5-Aminotetralin

Formula: C₁₀H₁₃N
Boiling Pt: 275 °C
Density: 1.06 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD00001736
CAS Number: 2217-41-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1aba342f8c0253047903d115846cf057181d4f61&vid=1aba342f8c0253047903d115846cf057181d4f61&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR