1-(Dimethylisopropylsilyl)imidazole ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: DMIPS1-(Isopropyldimethylsilyl)imidazole

D1596-1G D1596-5G
TCD1596-005GEA 740.85 CAD
TCD1596-005G TCD1596-001G
1-(Dimethylisopropylsilyl)imidazole ≥98.0% (by titrimetric analysis)
1-(Dimethylisopropylsilyl)imidazole

[Dimethylisopropylsilylating Agent]
CAS Number: 81452-04-2
MDL Number: MFCD00059891
Molecular Formula: C8H16N2Si
Molecular Weight: 168.32
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Storage Temperature: 0-10°C


Product Categories: Analytical Science, Silylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: DMIPS, 1-(Isopropyldimethylsilyl)imidazole

Formula: C₈H₁₆N₂Si
MW: 168.32 g/mol
MDL Number: MFCD00059891
CAS Number: 81452-04-2
https://ca.vwr.com/stibo/web/std.lang.all/83/97/28978397.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR