1,1'-Diheptyl-4,4'-bipyridinium dibromide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D1593-5G D1593-25G
TCD1593-005GEA 146.94 CAD
TCD1593-005G TCD1593-25G
1,1'-Diheptyl-4,4'-bipyridinium dibromide ≥98.0% (by HPLC, titration analysis)
1,1'-Diheptyl-4,4'-bipyridinium dibromide

[for Electrochromic Material]
CAS Number: 6159-05-3
MDL Number: MFCD00011997
Molecular Formula: C24H38Br2N2
Molecular Weight: 514.39
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal

Formula: C₂₄H₃₈Br₂N₂
MDL Number: MFCD00011997
CAS Number: 6159-05-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2cdaebd47980793c5ee08c5e43ce59b7e794e01&vid=e2cdaebd47980793c5ee08c5e43ce59b7e794e01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR