Dihydrolinalool ≥97.0%

Supplier: TCI America

Synonyms: 3,7-Dimethyl-6-octen-3-ol, 3,7-Dimethyloct-6-en-3-ol

D1592-500ML D1592-25ML
TCD1592-500MLEA 512.39 CAD
TCD1592-500ML TCD1592-025ML
Dihydrolinalool ≥97.0%
Dihydrolinalool

CAS Number: 18479-51-1
MDL Number: MFCD00059140
Molecular Formula: C10H20O
Molecular Weight: 156.27
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 200
Melting point (°C): 5
Flash Point (°C): 178
Specific Gravity (20/20): 0.86


Product Categories: Bioscience, Acyclic Monoterpenes
Other Product Names / Synonyms: 3,7-Dimethyl-6-octen-3-ol

Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 200 °C
Melting Pt: 5 °C
Density: 0.86 (20 °C)
Flash Pt: 178 °C
MDL Number: MFCD00059140
CAS Number: 18479-51-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aeae0f3303ee154f6979bac945b0b8d5c6f95c08&vid=aeae0f3303ee154f6979bac945b0b8d5c6f95c08&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR