(-)-1,4-Dithio-L-threitol ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: L-Cleland's Reagent, L-threo-1,4-Dimercaptobutane-2,3-diol, (2R,3R)-1,4-Dimercaptobutane-2,3-diol, L-Dithiothreitol, L-DTT, [R-(R*,R*)]-1,4-Dimercapto-2,3-butandiol, (2R,3R)-(-)-1,4-Dimercapto-2,3-butanediol, (2R,3R)-(-)-2,3-Dihydroxy-1,4-butanedithiol

D1589-5G D1589-1G
TCD1589-005GEA 1600.54 CAD
TCD1589-005G TCD1589-001G
(-)-1,4-Dithio-L-threitol ≥95.0% (by titrimetric analysis)
(-)-1,4-Dithio-L-threitol

CAS Number: 16096-97-2
MDL Number: MFCD00064305
Molecular Formula: C4H10O2S2
Molecular Weight: 154.24
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 51
Flash Point (°C): 110
Specific rotation [a]20/D: 26 deg (C=2.5, MeOH)
Storage Temperature: <0°C


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, Sugar Alcohols, Sugars (Others), O-Substituted Sugars
Other Product Names / Synonyms: L-Cleland's Reagent, (2R,3R)-(-)-2,3-Dihydroxy-1,4-butanedithiol, (2R,3R)-(-)-1,4-Dimercapto-2,3-butanediol, L-DTT

Formula: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Melting Pt: 51 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00064305
CAS Number: 16096-97-2
EINECS: 240-263-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0928a6c002e9d4059a004c47187253096555f125&vid=0928a6c002e9d4059a004c47187253096555f125&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR