(+)-Dehydroabietylamine ≥90.0%
Supplier: TCI America
D1588-5G
D1588-25G
TCD1588-005GEA
171.54
CAD
TCD1588-005G
TCD1588-025G
(+)-Dehydroabietylamine ≥90.0%
(+)-Dehydroabietylamine
[Optical Resolving Agent]
CAS Number: 1446-61-3
MDL Number: MFCD00213430
Molecular Formula: C20H31N
Molecular Weight: 285.48
Purity/Analysis Method: >90.0% (GC)
Melting point (°C): 40
Flash Point (°C): 110
Specific rotation [a]20/D: 45 deg (C=4, MeOH)
Formula:
C₂₀H₃₁N Melting Pt: 40 °C Flash Pt: 110 °C |
MDL Number:
MFCD00213430 CAS Number: 1446-61-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...