Dibutyl-N,N-diethylcarbamoylmethylphosphonate

Supplier: TCI America

Synonyms: N,N-Diethylcarbamoylmethylphosphonic Acid Dibutyl Ester

D1585-5ML D1585-25ML
TCD1585-005MLEA 83.74 CAD
TCD1585-005ML TCD1585-025ML
Dibutyl-N,N-diethylcarbamoylmethylphosphonate
Dibutyl-N,N-diethylcarbamoylmethylphosphonate

[for Extraction of Lanthanides and Actinides]
CAS Number: 7439-68-1
MDL Number: MFCD00015240
Molecular Formula: C14H30NO4P
Molecular Weight: 307.37
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 172
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: N,N-Diethylcarbamoylmethylphosphonic Acid Dibutyl Ester

Formula: C₁₄H₃₀NO₄P
MW: 307.37 g/mol
Boiling Pt: 172 °C
Density: 1.03 (20 °C)
MDL Number: MFCD00015240
CAS Number: 7439-68-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7297b09f48f70a63c6dec46ae30a566fa2495d89&vid=7297b09f48f70a63c6dec46ae30a566fa2495d89&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR