4'-(Dimethylamino)acetophenone ≥97.0% (by titrimetric analysis)

Supplier: TCI America

D1575-5G D1575-25G
TCD1575-005GEA 52.14 CAD
TCD1575-005G TCD1575-025G
4'-(Dimethylamino)acetophenone ≥97.0% (by titrimetric analysis)
4'-(Dimethylamino)acetophenone

CAS Number: 2124-31-4
MDL Number: MFCD00017246
Molecular Formula: C10H13NO
Molecular Weight: 163.22
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 105

Formula: C₁₀H₁₃NO
Melting Pt: 105 °C
MDL Number: MFCD00017246
CAS Number: 2124-31-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60034a2ed685993e7b5b37f2363c05c8449cf7c6&vid=60034a2ed685993e7b5b37f2363c05c8449cf7c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR