2,5-Dimethyl-4-hydroxy-3(2H)-furanone ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
4-Hydroxy-2,5-dimethyl-3(2H)-furanone, Furaneol, 4-Hydroxy-2,5-dimethylfuran-3(2H)-one
D1569-5G
D1569-25G
TCD1569-005GEA
66.36
CAD
TCD1569-005G
TCD1569-025G
2,5-Dimethyl-4-hydroxy-3(2H)-furanone ≥98.0% (by HPLC)
2,5-Dimethyl-4-hydroxy-3(2H)-furanone
CAS Number: 3658-77-3
MDL Number: MFCD00010706
Molecular Formula: C6H8O3
Molecular Weight: 128.13
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 81
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Furaneol, 4-Hydroxy-2,5-dimethyl-3(2H)-furanone
Formula:
C₆H₈O₃ MW: 128.13 g/mol Melting Pt: 81 °C |
MDL Number:
MFCD00010706 CAS Number: 3658-77-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...