Sodium dodecylbenzenesulfonate (mixture) ≥95.0% (by titrimetric analysis), soft type

Supplier: TCI America

D1566-25G D1566-5G
TCD1566-25GEA 619.83 CAD
TCD1566-25G TCD1566-005G
Sodium dodecylbenzenesulfonate (mixture) ≥95.0% (by titrimetric analysis), soft type
Sodium dodecylbenzenesulfonate (mixture)

CAS Number: 69669-44-9
MDL Number: MFCD00011508
Molecular Weight: 348.48
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Flash Point (°C): 110


Other Product Names / Synonyms: Dodecylbenzenesulfonic Acid Sodium Salt, Laurylbenzenesulfonic Acid Sodium Salt, Sodium Laurylbenzenesulfonate, Alkylbenzenesulfonic Acid Sodium Salt, Sodium Alkylbenzenesulfonate

Formula: C₁₈H₃₀O₃S
Flash Pt: 110 °C
MDL Number: MFCD00011508
CAS Number: 69669-44-9

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR