4-(3,5-Dibromo-2-pyridylazo)-1,3-phenylenediamine ≥98.0%

Supplier: TCI America

D1552-1G D1552-100MG
TCD1552-001GEA 456.15 CAD
TCD1552-001G TCD1552-100MG
4-(3,5-Dibromo-2-pyridylazo)-1,3-phenylenediamine ≥98.0%
4-(3,5-Dibromo-2-pyridylazo)-1,3-phenylenediamine

[for Colorimetric Analysis of Co, Cd]
CAS Number: 50768-79-1
MDL Number: MFCD00059780
Molecular Formula: C11H9Br2N5
Molecular Weight: 371.04
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₁₁H₉Br₂N₅
MW: 371.04 g/mol
MDL Number: MFCD00059780
CAS Number: 50768-79-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4045552dae73983d815f5abae141386dd47b9c9d&vid=4045552dae73983d815f5abae141386dd47b9c9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR