2,3-Dihydro-2-methylbenzofuran ≥98.0%

Supplier: TCI America

D1537-25G D1537-10G
TCD1537-25GEA 722.06 CAD
TCD1537-25G TCD1537-010G
2,3-Dihydro-2-methylbenzofuran ≥98.0%
2,3-Dihydro-2-methylbenzofuran

CAS Number: 1746-11-8
MDL Number: MFCD00023003
Molecular Formula: C9H10O
Molecular Weight: 134.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 198
Flash Point (°C): 63
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: 2-Methyl-2,3-dihydrobenzofuran

Formula: C₉H₁₀O
Boiling Pt: 198 °C
Density: 1.03 (20 °C)
Flash Pt: 63 °C
MDL Number: MFCD00023003
CAS Number: 1746-11-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9de23f93e577bf347ba11d458224696c9b02367d&vid=9de23f93e577bf347ba11d458224696c9b02367d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR