Diethylene glycol monomethyl ether methacrylate ≥97.0% stabilised

Supplier: TCI America

Synonyms: Methacrylic Acid 2-(2-Methoxyethoxy)ethyl Ester, 2-(2-Methoxyethoxy)ethyl Methacrylate, Methyl Carbitol Methacrylate

D1531-25ML D1531-500ML
TCD1531-025MLEA 42.45 CAD
TCD1531-025ML TCD1531-500ML
Diethylene glycol monomethyl ether methacrylate ≥97.0% stabilised
Diethylene glycol monomethyl ether methacrylate

(stabilized with MEHQ)
CAS Number: 45103-58-0
MDL Number: MFCD00059330
Molecular Formula: C9H16O4
Molecular Weight: 188.22
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 111
Specific Gravity (20/20): 1.02
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Methacrylic Acid 2-(2-Methoxyethoxy)ethyl Ester, 2-(2-Methoxyethoxy)ethyl Methacrylate, Methyl Carbitol Methacrylate

Formula: C₉H₁₆O₄
MW: 188.22 g/mol
Density: 1.02 (20 °C)
Flash Pt: 111 °C
MDL Number: MFCD00059330
CAS Number: 45103-58-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=30a9950063ef626d65168afef2534f53a338cdfe&vid=30a9950063ef626d65168afef2534f53a338cdfe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR