Diethylene glycol dimethacrylate ≥97.0% stabilized
Supplier: TCI America
D1530-25ML
D1530-500ML
D1530-100ML
TCD1530-025MLEA
26.86
CAD
TCD1530-025ML
TCD1530-500ML
TCD1530-100ML
Diethylene glycol dimethacrylate ≥97.0% stabilized
Diethylene glycol dimethacrylate
(stabilized with MEHQ)
CAS Number: 2358-84-1
MDL Number: MFCD00014931
Molecular Formula: C12H18O5
Molecular Weight: 242.27
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 134
Flash Point (°C): 140
Specific Gravity (20/20): 1.07
Formula:
C₁₂H₁₈O₅ Boiling Pt: 134 °C Density: 1.07 (20 °C) Flash Pt: 140 °C |
MDL Number:
MFCD00014931 CAS Number: 2358-84-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...