Diethylene glycol dibenzoate ≥97.0%
Supplier: TCI America
D1522-500ML
D1522-25ML
TCD1522-500MLEA
364.98
CAD
TCD1522-500ML
TCD1522-025ML
Diethylene glycol dibenzoate ≥97.0%
Diethylene glycol dibenzoate
CAS Number: 120-55-8
MDL Number: MFCD00020679
Molecular Formula: C18H18O5
Molecular Weight: 314.34
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 236
Flash Point (°C): 228
Freezing point (°C): 13
Specific Gravity (20/20): 1.18
Formula:
C₁₈H₁₈O₅ Boiling Pt: 236 °C Density: 1.18 (20 °C) Flash Pt: 228 °C |
MDL Number:
MFCD00020679 CAS Number: 120-55-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...