1-(Dimethylethylsilyl)imidazole ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D1516-1G D1516-5G
TCD1516-001GEA 123.24 CAD
TCD1516-001G TCD1516-005G
1-(Dimethylethylsilyl)imidazole ≥98.0% (by titrimetric analysis)
1-(Dimethylethylsilyl)imidazole

[Dimethylethylsilylating Agent]
CAS Number: 62365-34-8
MDL Number: MFCD00059723
Molecular Formula: C7H14N2Si
Molecular Weight: 154.29
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 80

Formula: C₇H₁₄N₂Si
Boiling Pt: 80 °C
MDL Number: MFCD00059723
CAS Number: 62365-34-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=329bb7c6176091b075a5ef02aa51cfe1c8ee453c&vid=329bb7c6176091b075a5ef02aa51cfe1c8ee453c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR