Dansyl-L-valine ≥98.0%

Supplier: TCI America

Synonyms: 5-Dimethylaminonaphthalene-1-sulfonyl-L-valine, Dns-Val-OH, 5-Dimethylaminonaphthalene-1-sulphonyl-L-valine

D1505-1G D1505-100MG
TCD1505-001GEA 428.18 CAD
TCD1505-001G TCD1505-100MG
Dansyl-L-valine ≥98.0%
Dansyl-L-valine

CAS Number: 1098-50-6
MDL Number: MFCD00069524
Molecular Formula: C17H22N2O4S
Molecular Weight: 350.43
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 140
Specific rotation [a]20/D: 19.5 deg (C=1, MeOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 5-Dimethylaminonaphthalene-1-sulfonyl-L-valine, Dns-Val-OH

Formula: C₁₇H₂₂N₂O₄S
MW: 350.43 g/mol
Melting Pt: 140 °C
MDL Number: MFCD00069524
CAS Number: 1098-50-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3647f6fa8d8fcd6003385ff7d779d119c3257764&vid=3647f6fa8d8fcd6003385ff7d779d119c3257764&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR