Dansylglycine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Dns-Gly-OH, N-[5-(Dimethylamino)naphthalene-1-sulfonyl]glycine, N-[5-(Dimethylamino)naphthalene-1-sulphonyl]glycine, 2-(5-(Dimethylamino)naphthalene-1-sulfonamido)acetic acid, 2-(5-(Dimethylamino)naphthalene-1-sulphonamido)acetic acid, Dansylglycine [for Albumin binding assay]

D1501-1G D1501-5G D1501-100MG
TCD1501-001GEA 243.32 CAD
TCD1501-001G TCD1501-5G TCD1501-100MG
Dansylglycine ≥98.0% (by titrimetric analysis)
Dansylglycine

CAS Number: 1091-85-6
MDL Number: MFCD00037734
Molecular Formula: C14H16N2O4S
Molecular Weight: 308.35
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 159


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-[5-(Dimethylamino)naphthalene-1-sulfonyl]glycine, Dns-Gly-OH

Formula: C₁₄H₁₆N₂O₄S
MW: 308.35 g/mol
Melting Pt: 159 °C
MDL Number: MFCD00037734
CAS Number: 1091-85-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2783c9a85b28d97a7cb8ea5af8199e610df3afc&vid=b2783c9a85b28d97a7cb8ea5af8199e610df3afc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR