3-Methoxy-3-methylbutyl acetate ≥98.0%

Supplier: TCI America

Synonyms: Acetic Acid 3-Methoxy-3-methylbutyl Ester

A0964-500ML A0964-25ML
TCA0964-500MLEA 147.1 CAD
TCA0964-500ML TCA0964-025ML
3-Methoxy-3-methylbutyl acetate ≥98.0%
3-Methoxy-3-methylbutyl acetate

CAS Number: 103429-90-9
MDL Number: MFCD00059340
Molecular Formula: C8H16O3
Molecular Weight: 160.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 74
Flash Point (°C): 73
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: Acetic Acid 3-Methoxy-3-methylbutyl Ester

Formula: C₈H₁₆O₃
MW: 160.21 g/mol
Boiling Pt: 74 °C
Density: 0.96 (20 °C)
Flash Pt: 73 °C
MDL Number: MFCD00059340
CAS Number: 103429-90-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3c4dbd28cb06fd9b17f1aab8caeda2f4e8d2a2f&vid=c3c4dbd28cb06fd9b17f1aab8caeda2f4e8d2a2f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR