Dextran, M.W. ~ 70,000

Supplier: TCI America

D1449-500G D1449-25G D1449-100G
TCD1449-500GEA 1261.69 CAD
TCD1449-500G TCD1449-025G TCD1449-100G
Dextran, M.W. ~ 70,000
Dextran

CAS Number: 9004-54-0
MDL Number: MFCD00130935
Form: Crystal
Color: White
Specific rotation [a]20/D: 185 deg (C=6, H2O)


Product Categories: Materials Chemistry, Polymers, Bioscience, Polysaccharides, Natural Polymers (for DDS)

Formula: (C₆H₁₀O₅)n
MDL Number: MFCD00130935
CAS Number: 9004-54-0
EINECS: 232-677-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR