Dextran, M.W. ~ 40,000
Supplier: TCI America
D1448-500G
D1448-25G
D1448-100G
TCD1448-500GEA
925.88
CAD
TCD1448-500G
TCD1448-025G
TCD1448-100G
Dextran, M.W. ~ 40,000
Dextran
CAS Number: 9004-54-0
MDL Number: MFCD00130935
Form: Crystal
Color: White
Specific rotation [a]20/D: 197 deg (C=6, H2O)
Product Categories: Materials Chemistry, Polymers, Bioscience, Polysaccharides, Natural Polymers (for DDS)
Formula:
(C₆H₁₀O₅)n |
MDL Number:
MFCD00130935 CAS Number: 9004-54-0 EINECS: 232-677-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...