4-Allylveratrole ≥98.0% for synthesis

Supplier: TCI America
Warning

Synonyms: 4-Allylveratrole, Eugenol methyll ether, 4-Prop-2-enylveratrole, 4-Allyl-1,2-Dimethoxybenzene, 4-Allyl-1,2-dimethoxybenzene, Methyl eugenol, Eugenol methyl ether, Methyleugenol

D1360-500ML D1360-25ML
TCD1360-500MLEA 274.92 CAD
TCD1360-500ML TCD1360-025ML
4-Allylveratrole ≥98.0% for synthesis
4-Allylveratrole

CAS Number: 93-15-2
MDL Number: MFCD00008652
Molecular Formula: C11H14O2
Molecular Weight: 178.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Melting point (°C): -4
Flash Point (°C): 130
Specific Gravity (20/20): 1.04


Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: Eugenol Methyl Ether, Methyleugenol, 4-Allylveratrole

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 255
Melting Pt: -4
Density: 1.04 (20 °C)
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00008652
CAS Number: 93-15-2
EINECS: 202-223-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c733f2f3fe0bfbeffb460c04e39c2358b63c29d&vid=f99b67b4c4f49820c56c48f6a977b0c47edcf329&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR