Ethyl (2-amino-4-thiazolyl)acetate ≥98.0%
Supplier: TCI America
Synonyms:
(2-Amino-4-thiazolyl)acetic Acid Ethyl Ester, Ethyl 2-(2-aminothiazol-4-yl)acetate, Ethyl 2-amino-4-thiazoleacetate
A0955-25G
A0955-500G
TCA0955-025GEA
39.5
CAD
TCA0955-025G
TCA0955-500G
Ethyl (2-amino-4-thiazolyl)acetate ≥98.0%
Ethyl (2-amino-4-thiazolyl)acetate
CAS Number: 53266-94-7
MDL Number: MFCD00005330
Molecular Formula: C7H10N2O2S
Molecular Weight: 186.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 94
Other Product Names / Synonyms: (2-Amino-4-thiazolyl)acetic Acid Ethyl Ester
Formula:
C₇H₁₀N₂O₂S MW: 186.23 g/mol Melting Pt: 94 °C |
MDL Number:
MFCD00005330 CAS Number: 53266-94-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...