1,3-Diphenylguanidine sulfate

Supplier: TCI America

D1348-500G D1348-25G
TCD1348-500GEA 1314.56 CAD
TCD1348-500G TCD1348-025G
1,3-Diphenylguanidine sulfate
1,3-Diphenylguanidine sulfate

CAS Number: 32514-47-9
MDL Number: MFCD00060241
Molecular Formula: C13H13N3
Molecular Weight: 520.61
Form: Crystal
Color: White

Formula: C₁₃H₁₃N₃
MDL Number: MFCD00060241
CAS Number: 32514-47-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9f78292566ad129c3ceaf3b3ea08d350742e67e&vid=d9f78292566ad129c3ceaf3b3ea08d350742e67e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR