Sodium-2,4-dichlorophenoxyacetate monohydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Na-2,4-D, 2,4-Dichlorophenoxyacetic Acid Sodium Salt

D1319-25G D1319-500G
TCD1319-025GEA 79 CAD
TCD1319-025G TCD1319-500G
Sodium-2,4-dichlorophenoxyacetate monohydrate ≥98.0% (by HPLC, titration analysis)
Sodium-2,4-dichlorophenoxyacetate monohydrate

CAS Number: 2702-72-9
MDL Number: MFCD00068284
Molecular Formula: C8H6Cl2O3
Molecular Weight: 243.01
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 216


Product Categories: Bioscience, Auxins
Other Product Names / Synonyms: 2,4-Dichlorophenoxyacetic Acid Sodium Salt, Na-2,4-D

Formula: C₈H₆Cl₂O₃
MW: 243.01 g/mol
Melting Pt: 216 °C
MDL Number: MFCD00068284
CAS Number: 2702-72-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=700916c2b66f9e81f8531f018be6d770024832ab&vid=700916c2b66f9e81f8531f018be6d770024832ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR