4-Amino-2,3,5,6-tetrafluorobenzamide ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

A0949-5G A0949-1G
TCA0949-005GEA 229.1 CAD
TCA0949-005G TCA0949-001G
4-Amino-2,3,5,6-tetrafluorobenzamide ≥98.0% (by GC, total nitrogen)
4-Amino-2,3,5,6-tetrafluorobenzamide

CAS Number: 1548-74-9
MDL Number: MFCD00008000
Molecular Formula: C7H4F4N2O
Molecular Weight: 208.12
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Melting point (°C): 188

Formula: C₇H₄F₄N₂O
MW: 208.12 g/mol
Melting Pt: 188 °C
MDL Number: MFCD00008000
CAS Number: 1548-74-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae8d5337e19149cf9284e71a25914e098e2d8e01&vid=ae8d5337e19149cf9284e71a25914e098e2d8e01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR