1-(3-Dimethylaminophenyl)ethanol ≥98.0%

Supplier: TCI America

Synonyms: 1-(3-Dimethylaminophenyl)ethyl Alcohol, 3-Dimethylamino-alpha-methylbenzyl Alcohol

D1269-5G D1269-25G
TCD1269-5GEA 246.32 CAD
TCD1269-5G TCD1269-025G
1-(3-Dimethylaminophenyl)ethanol ≥98.0%
1-(3-Dimethylaminophenyl)ethanol

CAS Number: 5339-01-5
MDL Number: MFCD00059658
Molecular Formula: C10H15NO
Molecular Weight: 165.24
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 102
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: 3-Dimethylamino-alpha-methylbenzyl Alcohol, 1-(3-Dimethylaminophenyl)ethyl Alcohol

Formula: C₁₀H₁₅NO
Boiling Pt: 102 °C
Density: 1.04 (20 °C)
MDL Number: MFCD00059658
CAS Number: 5339-01-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=080e64b692d98ac94946f4992b81d326ba3d83ac&vid=080e64b692d98ac94946f4992b81d326ba3d83ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR