2,2'-Dithiodianiline (thulium ionophore I) ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D1246-25G D1246-500G
TCD1246-025GEA 241.74 CAD
TCD1246-025G TCD1246-500G
2,2'-Dithiodianiline (thulium ionophore I) ≥98.0% (by HPLC, titration analysis)
2,2'-Dithiodianiline (Thulium ionophore I)

CAS Number: 1141-88-4
MDL Number: MFCD00007703
Molecular Formula: C12H12N2S2
Molecular Weight: 248.36
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 92


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 2-Aminophenyl Disulfide, 2,2'-Diaminodiphenyl Disulfide, Di(2-aminophenyl) Disulfide

Formula: C₁₂H₁₂N₂S₂
Melting Pt: 92 °C
MDL Number: MFCD00007703
CAS Number: 1141-88-4
EINECS: 214-529-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=960b2617443a372c6207bdf25b374c96dd867a20&vid=960b2617443a372c6207bdf25b374c96dd867a20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR