Diethylene glycol diacetate ≥98.0%
Supplier: TCI America
D1230-25ML
D1230-500ML
TCD1230-025MLEA
69.52
CAD
TCD1230-025ML
TCD1230-500ML
Diethylene glycol diacetate ≥98.0%
Diethylene glycol diacetate
CAS Number: 628-68-2
MDL Number: MFCD00026211
Molecular Formula: C8H14O5
Molecular Weight: 190.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 250
Flash Point (°C): 135
Freezing point (°C): 19
Specific Gravity (20/20): 1.12
Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Oxybis(2-ethyl Acetate)
Formula:
C₈H₁₄O₅ Boiling Pt: 250 °C Density: 1.12 (20 °C) Flash Pt: 135 °C |
MDL Number:
MFCD00026211 CAS Number: 628-68-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...