Bis(3-pyridylmethyl)amine ≥97.0%
Supplier: TCI America
D1199-25ML
D1199-5ML
TCD1199-025MLEA
970.12
CAD
TCD1199-025ML
TCD1199-005ML
Bis(3-pyridylmethyl)amine ≥97.0%
Bis(3-pyridylmethyl)amine
CAS Number: 1656-94-6
MDL Number: MFCD00038043
Molecular Formula: C12H13N3
Molecular Weight: 199.26
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.12
Other Product Names / Synonyms: 3,3'-Dipicolylamine, alpha,alpha'-Iminodi(3-picoline)
Formula:
C₁₂H₁₃N₃ Density: 1.12 (20 °C) |
MDL Number:
MFCD00038043 CAS Number: 1656-94-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...