N,N'-Dicarbobenzoxy-L-cystine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D1197-5G D1197-1G
TCD1197-025GEA 550.69 CAD
TCD1197-025G TCD1197-5G TCD1197-001G
N,N'-Dicarbobenzoxy-L-cystine ≥98.0% (by titrimetric analysis)
N,N'-Dicarbobenzoxy-L-cystine

CAS Number: 6968-11-2
MDL Number: MFCD00022030
Molecular Formula: C22H24N2O8S2
Molecular Weight: 508.56
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 119
Specific rotation [a]20/D: -91 deg (C=8.8, AcOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N,N'-Di-Cbz-L-cystine, (Z-Cys-OH)2

Formula: C₂₂H₂₄N₂O₈S₂
Melting Pt: 119 °C
MDL Number: MFCD00022030
CAS Number: 6968-11-2

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR