Dehydroabietylamine (mixture of isomeres) 55.0%

Supplier: TCI America

D1180-25G D1180-500G
TCD1180-025GEA 54.97 CAD
TCD1180-025G TCD1180-500G
Dehydroabietylamine (mixture of isomeres) 55.0%
Dehydroabietylamine (mixture of isomeres)

MDL Number: MFCD00213430
Molecular Formula: C20H31N
Molecular Weight: 285.48
Purity/Analysis Method: >55.0% (GC)
Form: Clear Liquid
Boiling point (°C): 344
Flash Point (°C): 110
Specific Gravity (20/20): 1.00
Specific rotation [a]20/D: 40 deg (C=4, MeOH)

Formula: C₂₀H₃₁N
Boiling Pt: 344 °C
Density: 1.00 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00213430
CAS Number: 99306-87-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cfe6cd24d821de83dddf1c81dc80545c363f1d70&vid=cfe6cd24d821de83dddf1c81dc80545c363f1d70&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 55.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR