4,4'-Di-n-octyloxyazoxybenzene ≥95.0% (by total nitrogen basis)
Supplier: TCI America
D1123-5G
D1123-1G
TCD1123-005GEA
1147.08
CAD
TCD1123-005G
TCD1123-1G
4,4'-Di-n-octyloxyazoxybenzene ≥95.0% (by total nitrogen basis)
4,4'-Di-n-octyloxyazoxybenzene
CAS Number: 25729-12-8
MDL Number: MFCD00059445
Molecular Formula: C28H42N2O3
Molecular Weight: 454.66
Purity/Analysis Method: >95.0% (N)
Form: Crystal
Formula:
C₂₈H₄₂N₂O₃ |
MDL Number:
MFCD00059445 CAS Number: 25729-12-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...