2-Amino-2-thiazoline hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
A0936-25G
A0936-5G
TCA0936-025GEA
285.98
CAD
TCA0936-025G
TCA0936-5G
2-Amino-2-thiazoline hydrochloride ≥98.0% (by titrimetric analysis)
2-Amino-2-thiazoline hydrochloride
CAS Number: 3882-98-2
MDL Number: MFCD00012709
Molecular Formula: C3H6N2S
Molecular Weight: 138.61
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 200
Formula:
C₃H₆N₂S Melting Pt: 200 °C |
MDL Number:
MFCD00012709 CAS Number: 3882-98-2 EINECS: 217-224-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...