2,2'-Dipyridyl disulfide ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1,2-Di(pyridin-2-yl)disulphane, 1,2-Di(pyridin-2-yl)disulfane, 2,2'-Dithiodipyridine

D1114-250G D1114-25G D1114-5G
TCD1114-250GEA 1324.04 CAD
TCD1114-250G TCD1114-025G TCD1114-005G
2,2'-Dipyridyl disulfide ≥98.0% (by GC, titration analysis)
2,2'-Dipyridyl disulphide

[for Peptide Synthesis]
CAS Number: 2127-03-9
MDL Number: MFCD00006287
Molecular Formula: C10H8N2S2
Molecular Weight: 220.31
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 58


Product Categories: Synthetic Organic Chemistry, Disulfides, Condensation & Active Esterification, Peptide Synthesis, Bioscience, Functional Group Blocking Reagents
Other Product Names / Synonyms: 2,2'-Dithiodipyridine

Formula: C₁₀H₈N₂S₂
MW: 220.32 g/mol
Boiling Pt: 348
Melting Pt: 58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006287
CAS Number: 2127-03-9
EINECS: 218-343-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b35ec8c196508fb0597187766bc72a1b11f4b627&vid=b43395fb6d6716fe121b57a06458a4b120e9ff38&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR