2,2'-Dihydroxybenzophenone ≥99.0%
Supplier: TCI America
Synonyms:
Bis(2-hydroxyphenyl)methanone
D1099-10G
D1099-25G
TCD1099-010GEA
319.16
CAD
TCD1099-010G
TCD1099-25G
2,2'-Dihydroxybenzophenone ≥99.0%
2,2'-Dihydroxybenzophenone
CAS Number: 835-11-0
MDL Number: MFCD00002217
Molecular Formula: C13H10O3
Molecular Weight: 214.22
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 63
Formula:
C₁₃H₁₀O₃ MW: 214.2167 g/mol Melting Pt: 63 °C |
MDL Number:
MFCD00002217 CAS Number: 835-11-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...